Learn More
Thomas Scientific Tweezers for use with calibration weights
SDP
Catalog No. 501457377
Encompass
Accuris™ Instruments offers a range of precision calibration weights that are ideal for calibration of your Accuris lab balances. Each weight meets class F1 tolerance per OIML (International Organization of Legal Metrology) Recommendation R111. Made of highly polished, non-magnetic stainless steel, each calibration weight comes in a protective plastic case. The W1000 series of individual weights come in protective plastic cases with a screw top cover. The W1100-200 Weight Set includes 23 weights ranging from 1mg to 200g and is packaged in a protective carrying case with a set of tweezers, a dust brush and cleaning cloth. Special tweezers, with non-scratching plastic points are also available for proper handling; precision weights should not be touched with bare hands as oils and salts from the skin can stick to the surface of the weights. Class E2 Weight Set Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability.  Precisio
Catalog No. 50-145-7377 Supplier Thomas Scientific Supplier No. 1190B13
Accuris™ Instruments offers a range of precision calibration weights that are ideal for calibration of your Accuris lab balances. Each weight meets class F1 tolerance per OIML (International Organization of Legal Metrology) Recommendation R111. Made of highly polished, non-magnetic stainless steel, each calibration weight comes in a protective plastic case. The W1000 series of individual weights come in protective plastic cases with a screw top cover. The W1100-200 Weight Set includes 23 weights ranging from 1mg to 200g and is packaged in a protective carrying case with a set of tweezers, a dust brush and cleaning cloth. Special tweezers, with non-scratching plastic points are also available for proper handling; precision weights should not be touched with bare hands as oils and salts from the skin can stick to the surface of the weights. Class E2 Weight Set Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability.  Precisio