Learn More
World Precision Instrument Dumont Tweezers 5MS, 8.2cm, Straight, 0.09x0.05mm Tips
SDP
Catalog No. 50822807
Encompass
Dumostar Material Information
Highly resistant to metal fatigue, has great elasticity, and is durable (Rockwell Hardness measurement of 62). Resistant to mineral and organic acids, salt. Autoclavable. More likely to break than to bend. The maximum temperature this material can withstand is 550° C.
Catalog No. 50-822-807 Supplier World Precision Instrument Supplier No. 501764
Dumostar Material Information
Highly resistant to metal fatigue, has great elasticity, and is durable (Rockwell Hardness measurement of 62). Resistant to mineral and organic acids, salt. Autoclavable. More likely to break than to bend. The maximum temperature this material can withstand is 550° C.