Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Replacement Tip Set
SDP
Supplier:  Electron Microscopy Sciences 784102WFR
Encompass_Preferred
Tips are PEEK/CF30.
Tip Width: 6.6mm
Catalog No. 50-239-96