Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Replacement Tip Set
SDP
Catalog No. 5023990
Encompass_Preferred
Tips are PEEK/CF30.
Tip Width: 12 mm
Depth: 2.0 mm
Catalog No. 50-239-90 Supplier Electron Microscopy Sciences Supplier No. 784144WFR
Tips are PEEK/CF30.
Tip Width: 12 mm
Depth: 2.0 mm