Learn More
Ted Pella Inc PELCO Reverse, Curved
SDP
Catalog No. NC9007755
Encompass
High precision reverse action tweezers. Specially intended where a high precision self closing action is required, such as in microscopy and assembly applications. Extra fine tipped tweezers made of anti-acid / non-magnetic stainless steel.
Catalog No. NC9007755 Supplier Ted Pella Inc Supplier No. 5372NM
High precision reverse action tweezers. Specially intended where a high precision self closing action is required, such as in microscopy and assembly applications. Extra fine tipped tweezers made of anti-acid / non-magnetic stainless steel.