Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style 3M, 70 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7815401
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Pointed, thin and fine mini-tweezers.
OAL: 70 mm
Catalog No. 50-237-82