Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style 5, 110 mm OAL
SDP
Catalog No. 5023799
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Super-fine points.
OAL: 110 mm
Catalog No. 50-237-99 Supplier Electron Microscopy Sciences Supplier No. 7813812
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Super-fine points.
OAL: 110 mm