Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style H, 90 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7815001
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Short and wide with fine tips.
OAL: 90 mm
Catalog No. 50-237-85