Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style H, 90 mm OAL
SDP
Catalog No. 5023791
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Short and wide with fine tips.
OAL: 90 mm
Catalog No. 50-237-91 Supplier Electron Microscopy Sciences Supplier No. 7815012
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Short and wide with fine tips.
OAL: 90 mm