Learn More
Electron Microscopy Sciences Gatan 3View SEM Handling Tweezers
SDP
Catalog No. 501929107
Encompass
The Gatan 3View stub handling Tweezers are especially designed for handling the 2 mm thin pin of the 3View system stubs used in the Gatan 3View system. They are designed as side grippers for easy grabbing and holding of the 2 mm pin to facilitate handling and storing the 3View stubs. Length: 118 mm. Polsihed edges for good contact. Matte anti-glare finish. Serrated handles for extra grip. Low carbon anti-magnetic stainless steel. Swiss made.
Catalog No. 50-192-9107 Supplier Electron Microscopy Sciences Supplier No. 7595907
The Gatan 3View stub handling Tweezers are especially designed for handling the 2 mm thin pin of the 3View system stubs used in the Gatan 3View system. They are designed as side grippers for easy grabbing and holding of the 2 mm pin to facilitate handling and storing the 3View stubs. Length: 118 mm. Polsihed edges for good contact. Matte anti-glare finish. Serrated handles for extra grip. Low carbon anti-magnetic stainless steel. Swiss made.