Learn More
Electron Microscopy Sciences EMS Tweezers Style 0, Locking, High Precision, Flat edges, fine Tips
SDP
Catalog No. 501930563
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer with flat edges. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Flat edges, fine
OAL: 120 mm
Catalog No. 50-193-0563 Supplier Electron Microscopy Sciences Supplier No. 785110R
The tweezers are the Swiss highest quality. High precision tweezer with flat edges. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Flat edges, fine
OAL: 120 mm