Learn More
Electron Microscopy Sciences EMS Tweezers Style 1, Locking, High Precision, Strong, straight, fine Tips
SDP
Catalog No. 501930565
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision locking tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Strong, straight, fine
OAL: 120 mm
Catalog No. 50-193-0565 Supplier Electron Microscopy Sciences Supplier No. 785111R
The tweezers are the Swiss highest quality. High precision locking tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Strong, straight, fine
OAL: 120 mm