Learn More
Electron Microscopy Sciences EMS Tweezers Style 27, High Precision, Strong, sharp, fine Tips
SDP
Catalog No. 501930566
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Strong, sharp, fine
OAL: 135 mm
Catalog No. 50-193-0566 Supplier Electron Microscopy Sciences Supplier No. 7851127
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Strong, sharp, fine
OAL: 135 mm