Learn More
Electron Microscopy Sciences EMS Tweezers Style 5, Locking, High Precision, Extra fine bent with superior finish Tips
SDP
Catalog No. 501930573
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision locking tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Extra fine bent with superior finish
OAL: 110 mm
Catalog No. 50-193-0573 Supplier Electron Microscopy Sciences Supplier No. 783255BR
The tweezers are the Swiss highest quality. High precision locking tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Extra fine bent with superior finish
OAL: 110 mm