Learn More
Electron Microscopy Sciences EMS Tweezers Style 51S, High Precision, Extra fine, bent tips Tips
SDP
Catalog No. 501930572
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish. Useful for handling EM grids.
Material: SA Steel
Grade: High Precision
Tips: Extra fine, bent tips
OAL: 115 mm
Catalog No. 50-193-0572 Supplier Electron Microscopy Sciences Supplier No. 7832551S
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish. Useful for handling EM grids.
Material: SA Steel
Grade: High Precision
Tips: Extra fine, bent tips
OAL: 115 mm