Learn More
Electron Microscopy Sciences EMS Tweezers Style AC, High Precision, Strong Tips
SDP
Catalog No. 501930577
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish. Serrated grips.
Material: SA Steel
Grade: High Precision
Tips: Strong
OAL: 110 mm
Catalog No. 50-193-0577 Supplier Electron Microscopy Sciences Supplier No. 78325AC
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish. Serrated grips.
Material: SA Steel
Grade: High Precision
Tips: Strong
OAL: 110 mm