Learn More
Electron Microscopy Sciences EMS Tweezers Style F, High Precision, Straight, flat, squared head, thin, strong, superior finish Tips
SDP
Catalog No. 501930578
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Straight, flat, squared head, thin, strong, superior finish
OAL: 120 mm
Catalog No. 50-193-0578 Supplier Electron Microscopy Sciences Supplier No. 78325F
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel
Grade: High Precision
Tips: Straight, flat, squared head, thin, strong, superior finish
OAL: 120 mm