Learn More
Research Products International Corp Twelve Channel Reservoir, Polypropylene, 5 per Case
SDP
Catalog No. 501978516
Encompass
Twelve Channel Reservoir is ideal for multi-channel pipeting and automated workstations. Each reservoir has 22 ml capacity. Interior of channels feature conical design for minimum reagent waste. Polypropylene reservoir withstands most reagents and alpha numeric indexing allows quick channel identification. Packaged 5 per case.
Catalog No. 50-197-8516 Supplier Research Products International Corp Supplier No. 246235
Twelve Channel Reservoir is ideal for multi-channel pipeting and automated workstations. Each reservoir has 22 ml capacity. Interior of channels feature conical design for minimum reagent waste. Polypropylene reservoir withstands most reagents and alpha numeric indexing allows quick channel identification. Packaged 5 per case.