Learn More
Aviva Systems Biology Corp TWF1 Antibody - N-terminal region (ARP56487_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TWF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TWF1. Alternate names: A6, MGC23788, MGC41876, PTK9ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-61881