Learn More
Valutek™ Twill Pattern Long Staple Cotton Wiper, Cold Knife Cut
Catalog No. 191502026
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-150-2026 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2026 Supplier Valutek™ Supplier No. VTTPC99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% tightly woven long stable cotton. The fabric of this wiper is bias-cut and has a cross section of 120 x 60 threads per square inch. This wiper is clean and durable with high heat resistance.
  • 100% cotton
  • Cold knife cut edge
  • Excellent absorbency
  • High heat resistance enables high temperature tolerance
  • Compatible with IPA and other common solvents
  • All Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment.
  • The cotton wiper is ideal for high heat settings such as high purity welding
  • It is also commonly used for cleaning, polishing and burnishing magnetic media disk surfaces, and the cleanup of aqueous and organic solvent spills.
Specifications
Material Cotton
Edges Bias edge
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile