Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Twin-Blade Floor Squeegees

Catalog No. 17998603 Shop All Perfex Corp. Products
Change view
Click to view available options
Width (English):
18 in.
24 in.
30 in.
Color:
Black
Blue
Green
Orange
Purple
Red
Yellow
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Color
17-998-603 30 in. Yellow
17-998-583 18 in. Blue
17-998-584 18 in. Black
17-998-585 18 in. Green
17-998-586 18 in. Orange
17-998-587 18 in. Purple
17-998-588 18 in. Red
17-998-589 18 in. Yellow
17-998-590 24 in. Blue
17-998-591 24 in. Black
17-998-592 24 in. Green
17-998-593 24 in. Orange
17-998-594 24 in. Purple
17-998-595 24 in. Red
17-998-596 24 in. Yellow
17-998-597 30 in. Blue
17-998-598 30 in. Black
17-998-599 30 in. Green
17-998-600 30 in. Orange
17-998-601 30 in. Purple
17-998-602 30 in. Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 17-998-603 Supplier Perfex™ Supplier No. 9530Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Twin-blade PVC floor squeegees in 18, 24, and 30 in. widths with color options.

  • Dual-mounted PVC blades provide double the service life, one blade wipes the surface while the other acts as a splash guard.
  • Lightweight and durable squeegee with excellent tear-resistance. Non-marring, non-conductive PVC blades will not absorb bacteria, liquids, or odors. Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents.
  • Connects to the long variant of the Lite-N-Tite connectors only.
  • Available in 18 in., 24 in., and 30 in. blade widths. Eight color options are available to color-code equipment per facility zones. Including White, Red, Blue, Green, Yellow, Orange, Purple, and Black.

Specifications

Product Type Double-blade Squeegee
Color-coded Yes
Color Yellow
Material PVC
Width (English) 30 in.
Width (Metric) 76.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.