Learn More
Contec™ Twin Bucket
BIOPHARMAGREENER_CHOICE
Catalog No. 19168530 Shop All Contec Inc. Products
This item is not returnable. View return policy
Catalog No. 19-168-530 Supplier Contec™ Supplier No. 2743S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contec™ Twin Bucket is an autoclavable divided bucket with a curved stainless steel wringer.
  • Made of durable polypropylene.
  • Use one side of the bucket for fresh cleaning and disinfecting solutions, and the other for waste.
  • For added mobility, 4 casters are included
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Fully autoclavable (Except casters)
Specifications
Autoclavable Autoclavable