Learn More
World Precision Instrument Tinned Cu Alloy
SDP
Catalog No. 50822161
Encompass
Micro coaxial cables (MAXxxxx) are ideal for microelectrode fabrication and construction of similar research tools. The dual shielding eliminates electrical interference caused by radio frequencies (RF), electrostatic and microphonics (e.g., bending and vibration. Available with single or dual (twin) conductors.
Catalog No. 50-822-161 Supplier World Precision Instrument Supplier No. MAX4020
Add to Cart
Add to Cart
Micro coaxial cables (MAXxxxx) are ideal for microelectrode fabrication and construction of similar research tools. The dual shielding eliminates electrical interference caused by radio frequencies (RF), electrostatic and microphonics (e.g., bending and vibration. Available with single or dual (twin) conductors.