Learn More
Electron Microscopy Sciences Twin Jet Electropolisher Model 501b
SDP
Catalog No. 50996994
Encompass
The METALTHIN Twin-Jet Electropolisher was developed for producing high quality thin foils from radioactive materials. It is robust, compact, economical, and easy to use.
The electrolyte tank, sample holder, thinning cell and tank base stand are machined from solid PVC to provide maximum resistance to corrosive electrolytes. The Cell parts are held together by stainless steel screws so that the Cell is easy to disassemble.
Catalog No. 50-996-994 Supplier Electron Microscopy Sciences Supplier No. 55220501B
Add to Cart
Add to Cart
Request a Quote
The METALTHIN Twin-Jet Electropolisher was developed for producing high quality thin foils from radioactive materials. It is robust, compact, economical, and easy to use.
The electrolyte tank, sample holder, thinning cell and tank base stand are machined from solid PVC to provide maximum resistance to corrosive electrolytes. The Cell parts are held together by stainless steel screws so that the Cell is easy to disassemble.