Learn More
Med Vet International Twin-Point Fragment Forceps, Straight, 6.5in, J0212TS, Each
Catalog No. 5000585486
Encompass_Preferred
Single pointed reduction forceps typically apply pressure at the exact point where it is desirable to place an implant. Splitting the tips allows implants to be placed at the ideal position (e.g., repair of lateral condyle fractures).
Catalog No. 50-005-85486 Supplier Med Vet International Supplier No. JOR212TS
Single pointed reduction forceps typically apply pressure at the exact point where it is desirable to place an implant. Splitting the tips allows implants to be placed at the ideal position (e.g., repair of lateral condyle fractures).