Learn More
Electron Microscopy Sciences Twin Tip Teeny Writer, Black, Fine/Ultra Fine (0.7/0.3 mm)
SDP
Catalog No. 50996485
Encompass_Preferred
Teeny Writers are perfect general use lab markers. The original Teeny Writer has an extra fine tip. Both are ideal for writing on Tough-Tags and Tough-Spots. Use for labeling samples stored at room temperature and not exposed to chemicals and/or caustic agents. The Twin-Tip Teeny Writer has a fine tip (0.7) and an ultra fine tip (0.3).
Color: Black
Size: 0.7/0.3 mm
Catalog No. 50-996-485 Supplier Electron Microscopy Sciences Supplier No. 62054B
Teeny Writers are perfect general use lab markers. The original Teeny Writer has an extra fine tip. Both are ideal for writing on Tough-Tags and Tough-Spots. Use for labeling samples stored at room temperature and not exposed to chemicals and/or caustic agents. The Twin-Tip Teeny Writer has a fine tip (0.7) and an ultra fine tip (0.3).
Color: Black
Size: 0.7/0.3 mm