Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Twin Tone Bar Set
GREENER_CHOICESCI_ED

Catalog No. S160134 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160134 6 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160134 Supplier Supertek™ Supplier No. PH36152
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use this Supertek™ Twin Tone Bar Set to demonstrate resonance, sympathetic vibrations and beat phenomenon.

  • Effective product to experiment on!
  • Two precision-made bars are mounted on a single solid resonance cavity.
  • The cavity has two chambers with one bar tuned to 440 Hz, the other to 441 Hz.
  • Size: 13.78 L x 7 W x 4.13 H in.

Specifications

Grade 6 to Undergraduate
Format Kit
Includes Hammer and instructions
Languages English
Subject Physics - Sound and Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.