Learn More
Research Products International Corp Twin UV Lamp, 2x365 NM
SDP
Catalog No. 501977194
Encompass
Twin UV Lamps feature two UV bulbs for applications requiring added UV intensity. Lamps are lightweight and easy to handle. Rugged, scratch-resistant painted metal housing. Optional stand available for hands free viewing.
Catalog No. 50-197-7194 Supplier Research Products International Corp Supplier No. 353069
Add to Cart
Add to Cart
Twin UV Lamps feature two UV bulbs for applications requiring added UV intensity. Lamps are lightweight and easy to handle. Rugged, scratch-resistant painted metal housing. Optional stand available for hands free viewing.