Learn More
Chemglass Life Sciences Twist It Stoppers, White, Assortment Pack of 33
SDP
Catalog No. 501531534
Encompass_Preferred
Twist It Stoppers, White, Assortment Pack of 33. Three twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper. Soft, resilient, natural rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving. Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Note: #3 & #4 only have two holes. The Assortment Pack Consists of the Following Stoppers: 9 each Size # 3, 8 each Size # 4, 6 each Size # 5, 4 each Size # 6, 2 each Size # 7, 2 each Size # 8, 1 each Size # 9, 1 each Size # 10.
Catalog No. 50-153-1534 Supplier Chemglass Life Sciences Supplier No. CG302909
Twist It Stoppers, White, Assortment Pack of 33. Three twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper. Soft, resilient, natural rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving. Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Note: #3 & #4 only have two holes. The Assortment Pack Consists of the Following Stoppers: 9 each Size # 3, 8 each Size # 4, 6 each Size # 5, 4 each Size # 6, 2 each Size # 7, 2 each Size # 8, 1 each Size # 9, 1 each Size # 10.