Learn More
MSC 3M Disinfectant Cleaner Concentrate, EPA-Registered, Hospital Grade, 2 L, Twist 'n Fill System
Catalog No. 502839588
Encompass
3M HB Quat Disinfectant Cleaner Concentrate 25H is an EPA-registered, high-yield hospital-grade disinfectant designed for use with the 3M Twist 'n Fill Chemical Management System. This rinse-free concentrate cleans, deodorizes, and disinfects hard, non-porous surfaces, effectively targeting a broad spectrum of bacteria, fungi, and viruses.
  • EPA-registered for broad-spectrum efficacy
  • Effective against HIV-1, HBV, MRSA, VRE, and other microorganisms
  • High-yield concentrate provides significant solution volume
  • Rinse-free formulation simplifies cleaning processes
  • Cleans, deodorizes, and disinfects various hard surfaces
  • Designed for precise dilution with chemical management systems
  • Supplied in 2 L bottles for convenient dispensing
Catalog No. 50-283-9588 Supplier MSC Supplier No. 89473219
3M HB Quat Disinfectant Cleaner Concentrate 25H is an EPA-registered, high-yield hospital-grade disinfectant designed for use with the 3M Twist 'n Fill Chemical Management System. This rinse-free concentrate cleans, deodorizes, and disinfects hard, non-porous surfaces, effectively targeting a broad spectrum of bacteria, fungi, and viruses.
  • EPA-registered for broad-spectrum efficacy
  • Effective against HIV-1, HBV, MRSA, VRE, and other microorganisms
  • High-yield concentrate provides significant solution volume
  • Rinse-free formulation simplifies cleaning processes
  • Cleans, deodorizes, and disinfects various hard surfaces
  • Designed for precise dilution with chemical management systems
  • Supplied in 2 L bottles for convenient dispensing