Learn More
Research Products International Corp Twist Rack Work Station with 4 Modules
SDP
Catalog No. 501978260
Encompass
Twist-Rack is the most convenient workstation for every laboratory storage application. Innovative system includes four rack modules that interlock to form any desired configuration. Simply twist end lock modules together for holding tube sizes from 0.2ml up to 50ml, tube strip or a 96-well plate.
Rack module capacity:
2 x 50 ml
8 x 15 ml
12 x 10.5 - 2.0 ml
6 x 0.5 ml
48 x 0.2 ml
6 x 8 tube strips
Two interlocked modules accommodate 8 x 12 tube strips or one 96-well plate. Complete workstation includes four polypropylene rack modules; green, yellow, blue and red. Racks can be autoclaved and withstand temperatures from -80°C to +121°C.
Catalog No. 50-197-8260 Supplier Research Products International Corp Supplier No. 140450
Twist-Rack is the most convenient workstation for every laboratory storage application. Innovative system includes four rack modules that interlock to form any desired configuration. Simply twist end lock modules together for holding tube sizes from 0.2ml up to 50ml, tube strip or a 96-well plate.
Rack module capacity:
2 x 50 ml
8 x 15 ml
12 x 10.5 - 2.0 ml
6 x 0.5 ml
48 x 0.2 ml
6 x 8 tube strips
Two interlocked modules accommodate 8 x 12 tube strips or one 96-well plate. Complete workstation includes four polypropylene rack modules; green, yellow, blue and red. Racks can be autoclaved and withstand temperatures from -80°C to +121°C.