Learn More
Aviva Systems Biology Corp TWIST1 Antibody - C-terminal region (ARP37997_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37997_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TWIST1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TWIST1. Alternate names: SCS, ACS3, CRS1, BPES2, BPES3, TWIST, bHLHa38ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56482