Learn More
Glass Expansion Twister Spray Chamber with Helix CT, 50ml cyclonic, Borosilicate glass. Includes UFT-16-75 UniFit Connector (1 unit) and 70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Supplier:  Glass Expansion 208090578HE
Encompass_Preferred
Twister spray chamber, includes Helix CT locking screw & seal (1 pc), and UniFit with 1.6mm OD x 0.75mm ID x 700mm long tube (1 pc). Made from borosilicate glass, 50mL volume. Compatible with PE Elan 5/6/9000 & Optima 3000 Series.The Twister features a baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma. Resulting in a more robust plasma, making the Twister an excellent choice for applications requiring the best possible precision & for both high TDS or organic sample matrices.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include NC1943061 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6936