Learn More
Glass Expansion Twister Spray Chamber with Long Neck and Helix CT, 50ml cyclonic, Borosilicate glass. Includes UFT-2-700 UniFit Connector (1 unit) and70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Catalog No. 501906923
Encompass_Preferred
Twister spray chamber with long neck, includes Helix CT locking screw & seal (1 pc) & UniFit with 2.0mm OD x 0.86mm ID x 700mm long tube (1 pc). Made from borosilicate glass, 50mL volume. Compatible with Spectro Arcos & Genesis EOP Series.The Twister features a baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma. Resulting in a more robust plasma, making the Twister an excellent choice for applications requiring the best possible precision & for both high TDS or organic sample matrices.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include UFT-2-700 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6923 Supplier Glass Expansion Supplier No. 208090243HE
Twister spray chamber with long neck, includes Helix CT locking screw & seal (1 pc) & UniFit with 2.0mm OD x 0.86mm ID x 700mm long tube (1 pc). Made from borosilicate glass, 50mL volume. Compatible with Spectro Arcos & Genesis EOP Series.The Twister features a baffle, which efficiently removes larger droplets, producing a smaller mean droplet size entering the plasma. Resulting in a more robust plasma, making the Twister an excellent choice for applications requiring the best possible precision & for both high TDS or organic sample matrices.The unique Helix CT interface consists of two parts, a ConstantTorque Locking Screw & chemically inert PressFit ferrule. The main feature of the Helix CT is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume, inert seal of the nebulizer.Replacement parts include UFT-2-700 & NC2081825, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001