Learn More
Glass Expansion Twister Spray Chamber for IsoMist ROL V2
SDP
Catalog No. 502703176
Encompass_Preferred
Twister Spray Chamber for IsoMist ROL V2. Replacement encapsulated Twister spray chamber for ROL version IsoMist XR units.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3176 Supplier Glass Expansion Supplier No. 218093186
Twister Spray Chamber for IsoMist ROL V2. Replacement encapsulated Twister spray chamber for ROL version IsoMist XR units.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001