Learn More
Glass Expansion Twister TFE Spray Chamber with Helix CT, 50ml cyclonic, PTFE. Includes UFT-075 UniFit Connector (1 unit) and 70-803-1439 Helix CT locking screw with seal (1 unit)
SDP
Catalog No. 501906950
Encompass_Preferred
Twister PTFE spray chamber, includes Helix CT locking screw & seal (1 pc) & UniFit with 1.3mm OD x 0.75mm ID x 700mm long tube (1 pc). Made from PTFE for HF resistance, 50mL volume. Compatible with Agilent 5000 series and PE Avio 200/500 series ICP-OES.The Twister PTFE is a HF-resistant, baffled cyclonic design with a proprietary Stediflow surface treatment that significantly improves sensitivity & precision, in addition to enhancing washout efficiency compared to other inert spray chamber designs. The PTFE Twister is an excellent choice for high matrix samples with HF. The interior surface can be refurbished at a nominal cost.The main feature of the Helix CT interface is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume & chemically inert seal of the nebulizer.Replacement parts include UFT-075, NC2081825 & V-044, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6950 Supplier Glass Expansion Supplier No. 208093452
Twister PTFE spray chamber, includes Helix CT locking screw & seal (1 pc) & UniFit with 1.3mm OD x 0.75mm ID x 700mm long tube (1 pc). Made from PTFE for HF resistance, 50mL volume. Compatible with Agilent 5000 series and PE Avio 200/500 series ICP-OES.The Twister PTFE is a HF-resistant, baffled cyclonic design with a proprietary Stediflow surface treatment that significantly improves sensitivity & precision, in addition to enhancing washout efficiency compared to other inert spray chamber designs. The PTFE Twister is an excellent choice for high matrix samples with HF. The interior surface can be refurbished at a nominal cost.The main feature of the Helix CT interface is the built-in, ratcheting torque control mechanism that allows for a consistent, zero dead volume & chemically inert seal of the nebulizer.Replacement parts include UFT-075, NC2081825 & V-044, suggested to replace every 6 months.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001