Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Twistit: Four Stoppers in One
SDPSCI_ED

Catalog No. S3508D Shop All Plasticoid Products
Change view
Click to view available options
Stopper Number:
5
6
7
Assorted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
S3508D 7
S3508C 6
S3508B 5
S35081 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S3508D Supplier Plasticoid Supplier No. J7M669 WHITE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-purpose and economical

Four stoppers in one! Seals twist off to adapt the stopper to your application. Use as a solid, one-, two-, or three-hole stopper. Reduces inventory, confusion, and cost. Made of clean, white, resilient, natural rubber with a clinical finish.

Specifications

Product Type Twistit Stopper
Stopper Number 7
Diameter (Metric) Bottom 3 cm
Diameter (Metric) Top 3.7 cm
Size 7
Total Positions 3
Description Clinical finish
Color White
Material Rubber
For Use With (Application) Multi-purpose applications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.