Learn More
Cole-Parmer Cole-Parmer Twistit white rubber stopper, size 6
Supplier:  Cole-Parmer 6299224
Encompass_Preferred
Use these versatile Twistit stoppers as either a solid, one-hole, two-hole, or three-hole stopper. Simply twist off a bottom seal to open a hole. The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Available in black or white rubber. Autoclavable.
Catalog No. NC1480278