Learn More
Cole-Parmer Two-Blade Folding Impeller, 5/16" Bore; 2" Dia
Supplier:  Cole-Parmer 0454300
Encompass
Lightnin 800578 Two-Blade Folding Impeller, 5/16" Bore; 2" Dia, 316 Stainless Steel, Two-blade folding impellers are perfect for mixing in vessels with narrow ports. Folded diameter is 11/16" for the 2" model and 1" for the 3" model. Impellers open fully when mixing.
Catalog No. NC1208907