Learn More
Waters Corp Cuvette Set, Two, Quartz, 190 to 800 nm Optical Range
Catalog No. 502392072 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This set includes two cuvettes, each made of high-quality quartz. They are designed for applications requiring an optical range of 190 to 800 nm, ensuring consistent performance for various spectroscopic measurements.
  • Two cuvettes included per set
  • Made of quartz material
  • Optical range of 190 to 800 nm
  • Both cuvettes sourced from the same lot of quartz for consistency
Catalog No. 50-239-2072 Supplier Waters Corp Supplier No. 700004155
This set includes two cuvettes, each made of high-quality quartz. They are designed for applications requiring an optical range of 190 to 800 nm, ensuring consistent performance for various spectroscopic measurements.
  • Two cuvettes included per set
  • Made of quartz material
  • Optical range of 190 to 800 nm
  • Both cuvettes sourced from the same lot of quartz for consistency