Learn More
Andwin Scientific AtmosBag, Two-hand, non-sterile, size M, closure type, Zipper-lock (39 in. × 48 in. × 15 in)
SDP
Catalog No. NC9431824
Encompass_Preferred
AtmosBag glove bag is a flexible, inflatable polyethylene chamber with built-in gloves that lets you work in a totally isolated and controlled environment. It is an inexpensive, portable alternative to the glove box and is well suited to a variety of tasks and materials. Specifications:• Constructed of sturdy 0.003 in. gauge polyethylene that is polished for good clarity• Seams are heat-sealed for strength and inflation tested to be leak-free• Inlet ports are provided for laboratory gas, vacuum, and electrical lines• AtmosBag is not fire-retardant or intended for prolonged contact with solvents, vapors, or chemicals• Two ports on each bag. One at the top and one on the side of the bag• Can be used in an Argon environment. 39 in. × 48 in. × 15 in., average inflated dimensions. 38 in. × 51.5 in. , deflated dimensions. Gas Volume 280 L
Catalog No. NC9431824 Supplier Andwin Scientific Supplier No. Z530212
AtmosBag glove bag is a flexible, inflatable polyethylene chamber with built-in gloves that lets you work in a totally isolated and controlled environment. It is an inexpensive, portable alternative to the glove box and is well suited to a variety of tasks and materials. Specifications:• Constructed of sturdy 0.003 in. gauge polyethylene that is polished for good clarity• Seams are heat-sealed for strength and inflation tested to be leak-free• Inlet ports are provided for laboratory gas, vacuum, and electrical lines• AtmosBag is not fire-retardant or intended for prolonged contact with solvents, vapors, or chemicals• Two ports on each bag. One at the top and one on the side of the bag• Can be used in an Argon environment. 39 in. × 48 in. × 15 in., average inflated dimensions. 38 in. × 51.5 in. , deflated dimensions. Gas Volume 280 L