Learn More
Synthware™ Two-Piece Vacuum Trap
GREENER_CHOICESDP
Catalog No. 31501816 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
31-501-816 1 Ea.
1 options
Catalog No. 31-501-816 Supplier Synthware™ Supplier No. V381512
Add to Cart
Add to Cart
Two-Piece design with a 0 to 12 mm high vacuum valve shut off
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Trap
Material Borosilicate Glass
Quantity 1 Ea.