Learn More
Cleatech TWO-PORT ISOLATION GLOVEBOX
SDP
Catalog No. NC1758485
Encompass_Preferred
2100-2-A Two-Port Isolation Glovebox,non-vacuum, Clear Acrylic, 35inWx 24inDx 25inH, two Internal access doors with Stainless steel frame, Removable fully gasketed back wall, 8in Diameter glove ports with stainless steel clamp
Catalog No. NC1758485 Supplier Cleatech Supplier No. 21002A
2100-2-A Two-Port Isolation Glovebox,non-vacuum, Clear Acrylic, 35inWx 24inDx 25inH, two Internal access doors with Stainless steel frame, Removable fully gasketed back wall, 8in Diameter glove ports with stainless steel clamp