Learn More
Electron Microscopy Sciences Binocular Two Power Stereo Microscope, 10X and 40X
SDP
Supplier:  Electron Microscopy Sciences STF804
Encompass_Preferred
Dissection and observation of macro specimens is more enlightening with our Stereo microscope. The large stage allows ample room for sample manipulation but still fits in a storage cabinet. Their illuminated stands let you choose light from three directions - above, below or both, to help observe opaque or transparent samples. Solid, all metal focusing mechanisms will endure student use well beyond the five year warranty. Binocular with two powers of magnification.
Magnification: 10X and 40X
Catalog No. 50-343-78