Learn More
Diversified Spaces™ Two-Station Steel Workbench with Six Horizontal Locker Openings
SCI_ED
Catalog No. S42428 Shop All Diversified Spaces Products
Change view
Click to view available options
Color:
Black
Gray
Includes:
No vises
One vises
Two vises
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
S42428 Gray No vises
S42429 Black No vises
S42430 Black One vises
S42431 Black Two vises
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S42428 Supplier Diversified Spaces™ Supplier No. WB6A0V
Only null left
Add to Cart
Add to Cart
Single-faced heavy gauge steel unit with one section construction and reinforced side panels available with or without vises in gray or black base unit.
  • Base consists of single-faced 20 gauge steel unit, welded and riveted throughout
  • Features six horizontal locker openings which measure 53.34D x 45.72W x 25.4cm H (21 x 18 x 10 in.) dimension
  • 18-gauge Steel locker doors with air vents, spring hinges, padlock hasp and knockout plug for cylinder lock
  • 5.715 (2.25 in.) thick maple top
  • Steel lockers with gray baked-enamel finish
  • SEFA compliant and MAS Certified Green
Specifications
Product Type 2-Station Steel Workbench with 6 Horizontal Locker Openings
Includes No vises
Color Gray
Dimensions (D x W x H) 71.12 x 162.56 x 84.45 cm (28 x 64 x 33.25 in.)
Warranty Limited Lifetime
Material Metal