Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 2-Stop, Chem-Durance Bio, 1.52 mm ID; 6/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9632736/DEL
Encompass
The overall length of Chem-Durance Bio 96327 and 96328-series stop tubing is 16" (406 mm). The legacy Ismatec MHLL stop tubing was 381 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1371061