Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Two Stroke Petrol Engine Model
GREENER_CHOICESCI_ED

Catalog No. S81682 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81682 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81682 Supplier Supertek™ Supplier No. PH20471
Add to Cart
Add to Cart

Showing internal structure and operating principles of a simple piston air cooled two stroke engine

  • Internal structure and operating principles of a simple piston air cooled two stroke engine
  • Mounted on a polished wood base with schematic diagram
  • Overall height approximately 15 in.

Specifications

Grade 6 to 12
Age 10 to 18
Includes Instruction manual
Class Size 1 Class
Product Type Two Stroke Petrol Engine Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.