Learn More
OHAUS Two-Tier Microplate Foam Inserts
Catalog No. 01804407 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-407 Vortex Attachment
1 options
Catalog No. 01-804-407 Supplier OHAUS Supplier No. 83041392
Add to Cart
Add to Cart
High-density, double layered foam insert for prolonged shaking of a standard microplate to protect sample from heat-buildup.
  • Fits into the 6 in. (152 mm) Platform attachment 146-6005-00.
  • For use with the Vortex-Genie 2 mixers and Vortex-Genie Pulse.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie