Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Two-Tier Polypropylene Test Tube Racks
GSA_VA

Catalog No. 1480920 Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
24
40
72
Holds:
Tubes (10 to 13 mm)
Tubes (13 to 16 mm)
Tubes (16 to 20 mm)
Tubes (20 to 30 mm)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions Holds
14-809-20 40 Tubes (13 to 16 mm)
03-337-13 24 Tubes (20 to 30 mm)
14-809-21 40 Tubes (16 to 20 mm)
03-213 72 Tubes (10 to 13 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-809-20
Only null left
Add to Cart
Edge
Add to Cart

Fit most water baths for elevated temperature incubations.

Fit most water baths for elevated temperature incubations.

  • Just the right size for serum vials (10-13 mm diameter), RIA vials and T3/T4 reaction vials (13-16 and 16-20 mm diameter), scintillation vials (25-30 mm diameter), or any other vial or tube of same dimensions
  • Long-lasting and corrosion-resistant
  • End plates handles
  • Polypropylene; steam autoclavable at 121°C (250°F)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Autoclavable
Color White
Holds Tubes (13 to 16 mm)
Material Polypropylene
Positions 40
Shape Rectangular
Stackable No
Sterility Non-sterile
Dimensions (L x W x H) 10 x 4.37 x 3.5 in. (25.4 x 11 x 8.9 cm)
Length (Metric) 25.4 cm
Length (English) 10 in.
Width (English) 4.37 in.
Height (English) 3.5 in.
Diameter (Metric) Opening 16 mm
Product Type Tube Rack (2-tier)
Diameter (Metric) Inner 16 mm
Shelves 2 Tiers
Height (Metric) 8.9 cm
Array 10 x 4
Width (Metric) 11 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.