Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Two-way normally open solenoid pinch valve; 24 VDC, 1/8"" ID x 1/4"" OD tubing
Catalog No. NC1616587
Encompass
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.
Catalog No. NC1616587 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9830236
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.